Merck
Influence of conformation on conductance of biphenyl-dithiol single-molecule contacts.
Nano letters (2009-12-23)
Artem Mishchenko, David Vonlanthen, Velimir Meded, Marius Bürkle, Chen Li, Ilya V Pobelov, Alexei Bagrets, Janne K Viljas, Fabian Pauly, Ferdinand Evers, Marcel Mayor, Thomas Wandlowski
ABSTRACT
The conductance of a family of biphenyl-dithiol derivatives with conformationally fixed torsion angle was measured using the scanning tunneling microscopy (STM)-break-junction method. We found that it depends on the torsion angle phi between two phenyl rings; twisting the biphenyl system from flat (phi = 0 degrees ) to perpendicular (phi = 90 degrees ) decreased the conductance by a factor of 30. Detailed calculations of transport based on density functional theory and a two level model (TLM) support the experimentally obtained cos(2) phi correlation between the junction conductance G and the torsion angle phi. The TLM describes the pair of hybridizing highest occupied molecular orbital (HOMO) states on the phenyl rings and illustrates that the pi-pi coupling dominates the transport under "off-resonance" conditions where the HOMO levels are well separated from the Femi energy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%
Supelco
Toluene-3,4-dithiol, for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%