Merck
  • Porous coordination polymer with flexibility imparted by coordinatively changeable lithium ions on the pore surface.
Porous coordination polymer with flexibility imparted by coordinatively changeable lithium ions on the pore surface.
Inorganic chemistry (2010-01-07)
Lin-Hua Xie, Jian-Bin Lin, Xiao-Min Liu, Yu Wang, Wei-Xiong Zhang, Jie-Peng Zhang, Xiao-Ming Chen
ABSTRACT
Solvothermal reactions of equimolar zinc acetate, lithium acetate, and 1,3,5-benzenetricarboxylic acid (H(3)btc) in different mixed solvents yielded isostructural three-dimensional frameworks [LiZn(btc)(cG)].lG [cG and lG denote coordinated and lattice guests, respectively; cG = (nmp)(0.5)(H(2)O)(0.5), lG = (EtOH)(0.5) (1a); cG = H(2)O, lG = EtOH (1b); nmp = N-methyl-2-pyrrolidone] with one-dimensional channels occupied by guest molecules and solvent-coordinated, extrusive Li(+) ions. Thermogravimetry analyses and powder X-ray diffraction measurements revealed that both 1a and 1b can lose all lattice and coordinated guests to form a desolvated phase [LiZn(btc)] (MCF-27, 1) and almost retains the original framework structure. Gas adsorption measurements on 1 confirmed its permanent porosity but suggested a structural transformation from 1a/1b to 1. It is noteworthy that only 1a can undergo a single-crystal to single-crystal (SCSC) transformation into 1 upon desolvation. The crystal structure of 1 revealed that the Li(+) ions were retracted into the channel walls via complementary coordination to the carboxylate oxygen atoms in the framework rather than being exposed on the pore surface. Single-crystal X-ray diffraction analyses were also performed for N(2)- and CO(2)-loaded samples of 1, revealing that the framework remained unchanged when the gases were adsorbed. Although the gas molecules could not be modeled, the residue electrons inside the channels demonstrated that the retracted Li(+) ions still behave as the primary interacting site for CO(2) molecules. Nevertheless, solvent molecules such as H(2)O can readily compete with the framework oxygen atom to retrieve the extrusive Li(+) ions, accompanying the reverse structural transformation, i.e., from 1 to 1a/1b.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Trimesic acid, 95%