Merck
  • Flash nano-welding: investigation and control of the photothermal response of ultrathin bismuth sulfide nanowire films.
Flash nano-welding: investigation and control of the photothermal response of ultrathin bismuth sulfide nanowire films.
Advanced materials (Deerfield Beach, Fla.) (2010-06-12)
Jordan W Thomson, Gregor Lawson, Paul O'Brien, Richard Klenkler, Michael G Helander, Srebri Petrov, Zheng-Hong Lu, Nazir P Kherani, Alex Adronov, Geoffery Ozin
ABSTRACT
Ultrathin Bi₂S₃ nanowires undergo a pronounced photothermal response to irradiation from a commercial camera flash. Controlled nano-welding was shown by using single walled carbon nanotube mats as an electrically and thermally conductive substrate. The resulting welded nanowire film is denser and has significantly lower resistance than unflashed bilayer films.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bismuth(III) sulfide, 99%