Merck
Comparison of methods for assessing integrity of equine sperm membranes.
Theriogenology (2011-04-19)
M L Foster, C C Love, D D Varner, S P Brinsko, K Hinrichs, S Teague, K Lacaze, T L Blanchard
ABSTRACT
Sperm membrane integrity (SMI) is thought to be an important measure of stallion sperm quality. The objective was to compare three methods for evaluating SMI: flow cytometry using SYBR-14/propidium iodide (PI) stain; an automated cell counting device using PI stain; and eosin-nigrosin stain. Raw equine semen was subjected to various treatments containing 20 to 80% seminal plasma in extender, with differing sperm concentrations, to simulate spontaneous loss of SMI. The SMI was assessed immediately, and after 1 and 2 d of cooled storage. Agreement between methods was determined according to Bland-Altman methodology. Eosin-nigrosin staining yielded higher (2%) overall mean values for SMI than did flow cytometry. Flow cytometry yielded higher (6%) overall mean values for SMI than did the automated cell counter. As percentage of membrane-damaged sperm increased, agreement of SMI measurement between methods decreased. When semen contained 50-79% membrane-intact sperm, the 95% limits of agreement between SMI determined by flow cytometry and eosin-nigrosin staining were greater (range = -26.9 to 24.3%; i.e., a 51.2% span) than for SMI determined by flow cytometry and the automated cell counter (range = -3.1 to 17.0%; 20.1% span). When sperm populations contained <50% membrane-intact sperm, the 95% limits of agreement between SMI determined by flow cytometry and eosin-nigrosin staining were greater (range = -35.9 to 19.0%; 54.9% span) than for SMI determined by flow cytometry and the automated cell counter (range = -11.6 to 28.7%; 40.3% span). We concluded that eosin-nigrosin staining assessments of percent membrane-intact sperm agreed less with flow cytometry when <80% of sperm had intact membranes, whereas automated cell counter assessments of percent membrane-intact sperm agreed less with flow cytometry when <30% of sperm had intact membranes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Nigrosin, certified by the BSC
Sigma-Aldrich
Nigrosin Stain solution, suitable for microscopy
Sigma-Aldrich
Nigrosin water soluble, For use as a biological stain