Merck
Degradation mechanism of Methyl Orange by electrochemical process on RuO(x)-PdO/Ti electrode.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-04-22)
Lin Du, Jin Wu, Song Qin, Changwei Hu
ABSTRACT
The electrochemical degradation of Methyl Orange in 0.1 M NaCl solution over RuO(x)-PdO/Ti anode was investigated. Chemical oxygen demand (COD), ion chromatography (IC), Fourier Transform Infrared Spectroscopy (FTIR) and Gas chromatography-mass spectrometry (GC-MS) were employed to detect the intermediates formed during the electrochemical degradation. In the present reaction system, Methyl Orange could be effectively degraded. After 1 h treatment, the discoloration could reach 97.9% with COD removal of 57.6%. The results indicated that in the presence of chloride, the electrolysis was able to oxidise the dye with partial mineralisation of carbon, nitrogen and sulfur into CO(2), NO(-)(3) and SO(2-)(4), respectively. After 8 h electrolysis, 62% of sulfur contained in Methyl Orange was transformed to SO(4)(2-), and 17.6% of nitrogen changed to NO(3)(-). The intermediates during electroprocess were detected to be low molecular weight compounds, chlorinated compounds, derivatives of benzene and long chain alkanes. Based on these data, a possible degradation mechanism of Methyl Orange was proposed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Palladium(II) oxide, 99.97% trace metals basis