Merck
  • Crystal structure of lactose permease in complex with an affinity inactivator yields unique insight into sugar recognition.
Crystal structure of lactose permease in complex with an affinity inactivator yields unique insight into sugar recognition.
Proceedings of the National Academy of Sciences of the United States of America (2011-05-20)
Vincent Chaptal, Seunghyug Kwon, Michael R Sawaya, Lan Guan, H Ronald Kaback, Jeff Abramson
ABSTRACT
Lactose permease of Escherichia coli (LacY) with a single-Cys residue in place of A122 (helix IV) transports galactopyranosides and is specifically inactivated by methanethiosulfonyl-galactopyranosides (MTS-gal), which behave as unique suicide substrates. In order to study the mechanism of inactivation more precisely, we solved the structure of single-Cys122 LacY in complex with covalently bound MTS-gal. This structure exhibits an inward-facing conformation similar to that observed previously with a slight narrowing of the cytoplasmic cavity. MTS-gal is bound covalently, forming a disulfide bond with C122 and positioned between R144 and W151. E269, a residue essential for binding, coordinates the C-4 hydroxyl of the galactopyranoside moiety. The location of the sugar is in accord with many biochemical studies.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sodium methanethiosulfonate, 95%