Merck
  • Evaluation of the P-glycoprotein- and breast cancer resistance protein-mediated brain penetration of 11C-labeled topotecan using small-animal positron emission tomography.
Evaluation of the P-glycoprotein- and breast cancer resistance protein-mediated brain penetration of 11C-labeled topotecan using small-animal positron emission tomography.
Nuclear medicine and biology (2011-07-02)
Tomoteru Yamasaki, Masayuki Fujinaga, Kazunori Kawamura, Akiko Hatori, Joji Yui, Nobuki Nengaki, Masanao Ogawa, Yuichiro Yoshida, Hidekatsu Wakizaka, Kazuhiko Yanamoto, Toshimitsu Fukumura, Ming-Rong Zhang
ABSTRACT
Topotecan (TPT) is a camptothecin derivative and is an anticancer drug working as a topoisomerase-I-specific inhibitor. But TPT cannot penetrate through the blood-brain barrier. In this study, we synthesized a new positron emission tomography (PET) probe, [(11)C]TPT, to evaluate the P-glycoprotein (Pgp)- and breast cancer resistance protein (BCRP)-mediated brain penetration of [(11)C]TPT using small-animal PET. [(11)C]TPT was synthesized by the reaction of a desmethyl precursor with [(11)C]CH(3)I. In vitro study using [(11)C]TPT was carried out in MES-SA and doxorubicin-resistant MES-SA/Dx5 cells in the presence or absence of elacridar, a specific inhibitor for Pgp and BCRP. The biodistribution of [(11)C]TPT was determined using small-animal PET and the dissection method in mice. The transport of [(11)C]TPT to the extracellular side was determined in MES-SA/Dx5 cells exhibiting the expressions of Pgp and BCRP at high levels. This transport was inhibited by coincubation with elacridar. In Mdr1a/b(-/-)Bcrp1(-/-) mice, PET results indicated that the brain uptake of [(11)C]TPT was about two times higher than that in wild-type mice. Similarly, the brain penetration of [(11)C]TPT in wild-type mice was increased by treatment with elacridar. The radioactivity in the brain of elacridar-treated mice was maintained at a certain level after the injection of [(11)C]TPT, although the radioactivity in the blood decreased with time. We demonstrated the increase of brain penetration of [(11)C]TPT by deficiency and inhibition of Pgp and BCRP functions using small-animal PET in mice.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Elacridar, ≥98% (HPLC)