Merck
  • Molecularly imprinted microspheres as SPE sorbent for selective extraction of four Sudan dyes in catsup products.
Molecularly imprinted microspheres as SPE sorbent for selective extraction of four Sudan dyes in catsup products.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-09-09)
Fengxia Qiao, Yuru Geng, Changqing He, Yupei Wu, Pengyu Pan
ABSTRACT
A highly selective molecularly imprinted solid-phase extraction (MISPE) coupled with high performance liquid chromatography (HPLC) ultraviolet-visible detection was developed for the simultaneous isolation and determination of four Sudan dyes (I, II, III and IV) in catsup products. The novel molecularly imprinted microspheres (MIM) were synthesized by aqueous suspension polymerization using phenylamine and naphthol as template, which showed high affinity to Sudan dyes in aqueous solution. In order to develop a selective extraction protocol for simultaneous determination the four Sudan dyes from catsup products, the molecular recognition properties of MIM as a SPE sorbent were evaluated. Under the optimized condition, good linearity was obtained from 0.01 to 2.5 μg g(-1) (r(2)≥ 0.9990) with the relative standard deviations of less than 3.4%. This proposed MISPE-HPLC procedure eliminated the effect of template leakage on quantitative analysis and could be applied to direct determination of four Sudan dyes in complicated food samples.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sudan I, Dye content ≥95 %
Supelco
Sudan I, analytical standard