Merck
  • Construction of cyclopentanol derivatives via three-component coupling of silyl glyoxylates, acetylides, and nitroalkenes.
Construction of cyclopentanol derivatives via three-component coupling of silyl glyoxylates, acetylides, and nitroalkenes.
Organic letters (2012-01-13)
Gregory R Boyce, Shubin Liu, Jeffrey S Johnson
ABSTRACT
The three-component coupling of Mg acetylides, silyl glyoxylates, and nitroalkenes results in a highly diastereoselective Kuwajima-Reich/vinylogous Michael cascade that provides tetrasubstituted silyloxyallene products. The regio- and diastereoselectivity were studied using DFT calculations. These silyloxyallenes were converted to cyclopentenols and cyclopentitols via a unique Lewis acid assisted Henry cyclization. The alkene functionality present in the cyclopentanol products can be elaborated using diastereoselective ketohydroxylation reactions.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Cyclopentanol, 99%