Merck
Michelangelo's eye disease.
Medical hypotheses (2012-03-20)
P E Gallenga, Giampiero Neri, Ruggero D'Anastasio, Vito Enrico Pettorrossi, Emilio Alfieri, Luigi Capasso
ABSTRACT
Charged by the Pope Julius II for painting the Cappella Sistina in Rome (between 1508 and 1512), Michelangelo worked in an elevated scaffolding, in an anomalous position with dyes (including poisoning lead salts) and solvents (such as toxic turpentine) dripping on his face and continuously inhaling, in a dim environment illuminated only with oil lamps and candles, as he described himself and sketched in a sonet addressed to Giovanni da Pistoia. In 1510 he began suffering from eye disease: the main symptom was the necessity to elevate the document he was reading up to the level of his eyes. This defect disappeared few months after he finished painting his masterpiece. We hypothesize that the Michelangelo's eyes disease was a form of acquired and transitory nystagmus induced by the many hours he spent in up gaze, with a skew deviation, a form of ocular tilt reaction resulting from the impairment of spatial sensitivity (inversion illusion) due to the persistence of the artist's head in a horizontal position, looking upward.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Oil of turpentine, purified