Merck
  • Organocatalytic asymmetric conjugate addition of 3-monosubstituted oxindoles to (E)-1,4-diaryl-2-buten-1,4-diones: a strategy for the indirect enantioselective furanylation and pyrrolylation of 3-alkyloxindoles.
Organocatalytic asymmetric conjugate addition of 3-monosubstituted oxindoles to (E)-1,4-diaryl-2-buten-1,4-diones: a strategy for the indirect enantioselective furanylation and pyrrolylation of 3-alkyloxindoles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-04-14)
Yu-Hua Liao, Xiong-Li Liu, Zhi-Jun Wu, Xi-Lin Du, Xiao-Mei Zhang, Wei-Cheng Yuan
ABSTRACT
An asymmetric conjugate addition of 3-monosubstituted oxindoles to a range of (E)-1,4-diaryl-2-buten-1,4-diones, catalyzed by commercially available cinchonine, is described. This organocatalytic asymmetric reaction affords a broad range of 3,3'-disubstituted oxindoles that contain a 1,4-dicarbonyl moiety and vicinal quaternary and tertiary stereogenic centers in high-to-excellent yields (up to 98%), with excellent diastereomeric and moderate-to-high enantiomeric ratios (up to 99:1 and 95:5, respectively). Subsequently, cyclization of the 1,4-dicarbonyl moiety in the resultant Michael adducts under different Paal-Knorr conditions results in two new kinds of 3,3'-disubstituted oxindoles--3-furanyl- and 3-pyrrolyl-3-alkyl-oxindoles--in high yields and good enantioselectivities. Notably, the studies presented here sufficiently confirm that this two-step strategy of sequential conjugate addition/Paal-Knorr cyclization is not only an attractive method for the indirect enantioselective heteroarylation of 3-alkyloxindoles, but also opens up new avenues toward asymmetric synthesis of structurally diverse 3,3'-disubstituted oxindole derivatives.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
(+)-Cinchonine, 85%
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)