Merck
Nitrosation of aryl and heteroaryltrifluoroborates with nitrosonium tetrafluoroborate.
The Journal of organic chemistry (2012-04-25)
Gary A Molander, Livia N Cavalcanti
ABSTRACT
Organotrifluoroborates have emerged as an alternative to toxic and air- and moisture-sensitive organometallic species for the synthesis of functionalized aryl and heteroaryl compounds. It has been shown that the trifluoroborate moiety can be easily converted into a variety of different substituents in a late synthetic stage. In this paper, we disclose a mild, selective, and convenient method for the ipso-nitrosation of organotrifluoroborates using nitrosonium tetrafluoroborate (NOBF(4)). Aryl- and heteroaryltrifluoroborates were converted into the corresponding nitroso products in good to excellent yields. This method proved to be tolerant of a broad range of functional groups.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Nitrosyl tetrafluoroborate, 95%