Merck
Pharmacokinetic evaluation of zolmitriptan for the treatment of migraines.
Expert opinion on drug metabolism & toxicology (2012-07-06)
Luana Lionetto, Barbara Casolla, Fabiola Mastropietri, Lidia D'Alonzo, Andrea Negro, Maurizio Simmaco, Paolo Martelletti
ABSTRACT
Migraine is a multifactorial neurovascular disorder characterized by recurrent episodes of disabling pain attacks, accompanied with gastrointestinal, neurological systems dysfunction. The pharmacologic treatment of migraine is classically divided in the management of the acute attack and preventive strategies. Acute treatments consist of triptan, ergot, opioid, antiemetic and NSAIDs. This article discusses pharmacodynamics and pharmacokinetics of zolmitriptan . The data were obtained by searching the following keywords in MEDLINE: zolmitriptan, pharmacokinetics, pharmacodynamics, triptans, migraine, menstrual-related migraine, cluster headache, relatively to the period 1989 - 2012. Zolmitriptan has been considered effective treatment in the acute phase of migraine, menstrual-related migraine and cluster headache attacks. Pharmacokinetic parameters may vary as a consequence of gender differences, inter- and intra-subjects variability and delivery system. Zolmitriptan was developed with the aim of obtaining a lipophilic compound in order to be more rapidly absorbed and centrally active. Pharmacologically, pharmacokinetic parameters are responsible for its wide efficacy and the limited adverse effect profile.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Zolmitriptan, ≥98% (HPLC)