Merck
  • Metabolism of tanshinone IIA, cryptotanshinone and tanshinone I from Radix Salvia miltiorrhiza in zebrafish.
Metabolism of tanshinone IIA, cryptotanshinone and tanshinone I from Radix Salvia miltiorrhiza in zebrafish.
Molecules (Basel, Switzerland) (2012-07-20)
Yingjie Wei, Ping Li, Changmei Wang, Yunru Peng, Luan Shu, Xiaobin Jia, Wenquan Ma, Bing Wang
ABSTRACT
The study aimed to investigate the potential of zebrafish in imitating mammal phase I metabolism of natural compounds. Three diterpenoid quinones from Radix Salvia miltiorrhiza, namely tanshinone IIA (TIIA), cryptotanshinone (Cry) and tanshinone I (TI) were selected as model compounds, and their metabolites mediated by zebrafish were characterized using a high-performance liquid chromatography coupled ion-trap mass spectrometry (HPLC/IT-MSn) method with electrospray ionization in positive mode. The separation was performed with a Zorbax C-18 column using a binary gradient elution of 0.05% formic acid acetonitrile/0.05% formic acid water. According to the MS spectra and after comparison with reference standards and literature reports, hydroxylation, dehydrogenation or D-ring hydrolysis metabolites of TIIA and Cry but not of TI were characterized, which coincided with those reported using regular in vivo or in vitro metabolic analysis methods, thus verifying that zebrafish can successfully imitate mammalian phase I metabolism which instills further confidence in using zebrafish as a novel and prospective metabolism model.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Cryptotanshinone, ≥98% (HPLC)