Merck
  • Design, synthesis and biological evaluation of some novel thienopyrimidines and fused thienopyrimidines as anti-inflammatory agents.
Design, synthesis and biological evaluation of some novel thienopyrimidines and fused thienopyrimidines as anti-inflammatory agents.
European journal of medicinal chemistry (2012-07-28)
Ola H Rizk, Omaima G Shaaban, Ibrahim M El-Ashmawy
ABSTRACT
Some new substituted thienopyrimidine derivatives comprising thioxo, thioalkyl and pyrazolyl derivatives as well as fused thienotriazolopyrimidine and thienopyrimidinotriazine ring systems were prepared from 3-benzyl-2-hydrazino-5-methyl-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxamide 4. The designed compounds were evaluated for their anti-inflammatory activity. Compounds 4, 9, 10 and 13 showed the highest anti-inflammatory effect compared with the reference drug diclofenac sodium.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Oil of turpentine, purified