Merck
  • The histopathological comparison of L-carnitine with amifostine for protective efficacy on radiation-induced acute small intestinal toxicity.
The histopathological comparison of L-carnitine with amifostine for protective efficacy on radiation-induced acute small intestinal toxicity.
Journal of cancer research and therapeutics (2012-07-31)
Murat Caloglu, Vuslat Yurut Caloglu, Tulin Yalta, Omer Yalcin, Cem Uzal
ABSTRACT
The aim of the study was to compare the protective efficacy of l-carnitine (LC) to amifostine on radiation-induced acute small intestine damage. Thirty, 4-week-old Wistar albino rats were randomly assigned to four groups - Group 1: control (CONT, n = 6), Group 2: irradiation alone (RT, n = 8), Group 3: amifostine plus irradiation (AMI+RT, n = 8), and Group 4: l-Carnitine plus irradiation (LC+RT, n = 8). The rats in all groups were irradiated individually with a single dose of 20 Gy to the total abdomen, except those in CONT. LC (300 mg/kg) or amifostine (200 mg/kg) was used 30 min before irradiation. Histopathological analysis of small intestine was carried out after euthanasia. Pretreatment with amifostine reduced the radiation-induced acute degenerative damage (P = 0.009) compared to the RT group. Pretreatment with LC did not obtain any significant difference compared to the RT group. The vascular damage significantly reduced in both of the AMI+RT (P = 0.003) and LC+RT group (P = 0.029) compared to the RT group. The overall damage score was significantly lower in the AMI+RT group than the RT group (P = 0.009). There was not any significant difference between the LC+RT and RT group. Amifostine has a marked radioprotective effect against all histopathological changes on small intestinal tissue while LC has limited effects which are mainly on vascular structure.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Amifostine trihydrate, ≥97% (TLC), powder