Merck
  • The antimicrobial activity of alpha-bisabolol and tea tree oil against Solobacterium moorei, a Gram-positive bacterium associated with halitosis.
The antimicrobial activity of alpha-bisabolol and tea tree oil against Solobacterium moorei, a Gram-positive bacterium associated with halitosis.
Archives of oral biology (2012-09-04)
Marcel Forrer, Eva M Kulik, Andreas Filippi, Tuomas Waltimo
ABSTRACT
To investigate the antimicrobial effect of alpha-bisabolol and tea tree oil alone and in combination against the halitosis-associated Gram-positive bacillus Solobacterium moorei. The inhibitory activity of alpha-bisabolol and tea tree oil against the reference strain S. moorei CCUG39336 and four clinical S. moorei isolates was investigated by a direct exposure test. Additionally, the ability of alpha-bisabolol to increase the sensitivity of S. moorei was tested by pretreating the bacteria with sublethal concentrations prior to the administration of tea tree oil. A dose-dependent killing was observed for the antimicrobial agents in a direct exposure test with the reference strain S. moorei CCUG39336. Concentrations of ≥0.5% tea tree oil caused decreases in viability of >5 log colony forming units/ml even after short incubation periods, while bacterial viability was less affected by alpha-bisabolol. The combination of 0.1% alpha-bisabolol plus 0.05% tea tree oil showed a synergistic effect on S. moorei strain CCUG39336 and on two of the four clinical S. moorei isolates tested. However, incubation of S. moorei with a sublethal concentration of 0.1% alpha-bisabolol for three days prior to the administration of 0.05% tea tree oil did not enhance the antibacterial effect of tea tree oil. Halitosis-associated bacterium S. moorei is susceptible to the antimicrobial agents tea tree oil and alpha-bisabolol, suggesting that these compounds might be beneficial in oral healthcare products.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
(−)-α-Bisabolol, ≥93% (GC)
Sigma-Aldrich
Tea tree oil, FG
Supelco
(−)-α-Bisabolol, analytical standard
(−)-α-Bisabolol, primary reference standard