Merck
  • The effectiveness of a single-stage versus traditional three-staged protocol of hospital disinfection at eradicating vancomycin-resistant Enterococci from frequently touched surfaces.
The effectiveness of a single-stage versus traditional three-staged protocol of hospital disinfection at eradicating vancomycin-resistant Enterococci from frequently touched surfaces.
American journal of infection control (2012-09-18)
N Deborah Friedman, Aaron L Walton, Sarah Boyd, Christopher Tremonti, Jillian Low, Kaylene Styles, Owen Harris, David Alfredson, Eugene Athan
ABSTRACT
Environmental contamination is a reservoir for vancomycin-resistant enterococcus (VRE) in hospitals. Environmental sampling of surfaces was undertaken anytime before disinfection and 1 hour after disinfection utilizing a sodium dichloroisocyanurate-based, 3-staged protocol (phase 1) or benzalkonium chloride-based, single-stage clean (phase 2). VRE colonization and infection rates are presented from 2010 to 2011, and audits of cleaning completeness were also analyzed. Environmental samples collected before disinfection were significantly more likely to be contaminated with VRE during phase 1 than phase 2: 25.2% versus 4.6%, respectively; odds ratio (OR), 7.01 (P < .01). Environmental samples collected after disinfection were also significantly more likely to yield VRE during phase 1 compared with phase 2: 11.2% versus 1.1%, respectively; OR, 11.73 (P < .01). Rates of VRE colonization were higher during 2010 than 2011. Cleaning audits showed similar results over both time periods. During use of a chlorine-based, 3-staged protocol, significantly higher residual levels of VRE contamination were identified, compared with levels detected during use of a benzalkonium chloride-based product for disinfection. This reduction in VRE may be due to a new disinfection product, more attention to the thoroughness of cleaning, or other supplementary efforts in our institution.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sodium dichloroisocyanurate dihydrate, 98.0% (AT)
Sigma-Aldrich
Sodium dichloroisocyanurate, 96%