Merck
  • Synthesis and structural characterization of novel cyclam-based zirconium complexes and their use in the controlled ROP of rac-lactide: access to cyclam-functionalized polylactide materials.
Synthesis and structural characterization of novel cyclam-based zirconium complexes and their use in the controlled ROP of rac-lactide: access to cyclam-functionalized polylactide materials.
Dalton transactions (Cambridge, England : 2003) (2012-10-09)
Luis G Alves, Frédéric Hild, Rui F Munhá, Luis F Veiros, Samuel Dagorne, Ana M Martins
ABSTRACT
Novel Bn(2)Cyclam-based zirconium complexes of the type (Bn(2)Cyclam)Zr(X)(X') (3, X = X' = OPh; 4, X = X' = SPh; 5, X = Cl, X' = O(i)Pr) were synthesized in good yields via metathesis routes involving the reaction of the dichloro precursor (Bn(2)Cyclam)Zr(Cl)(2) and the appropriate lithium salts. The molecular structures of compounds 3, 4 and 5, as determined by X-ray crystallographic studies, all confirmed the effective chelation of the Bn(2)Cyclam ligand in a κ(4)-N(2)N(2)' fashion, with the hexa-coordinated Zr center adopting a trigonal prismatic geometry. Complexes 3-5 as well as the diisopropoxide derivative (Bn(2)Cyclam)Zr(O(i)Pr)(2) (2) were all found to initiate the ring-opening polymerization (ROP) of rac-lactide in a controlled manner, as deduced from SEC data and linear correlations between molecular weight numbers (M(n)) and monomer conversion as the ROP proceeds. While initiator 2 polymerizes rac-lactide to afford, as expected, an O(i)Pr-ester-end PLA, the ROP of rac-lactide by species 3 or 4 affords an unusual cyclam-end group PLA, as deduced from MALDI-TOF data. The bonding and the electronic structures of the dialkoxides 2 and 3 were assessed by DFT and their possible influence on the polymerization mechanism is discussed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,4,8,11-Tetraazacyclotetradecane, 98% (GC)
Prezzi e disponibilità al momento non sono disponibili