Merck
  • FoxA1 corrupts the antiandrogenic effect of bicalutamide but only weakly attenuates the effect of MDV3100 (Enzalutamide™).
FoxA1 corrupts the antiandrogenic effect of bicalutamide but only weakly attenuates the effect of MDV3100 (Enzalutamide™).
Molecular and cellular endocrinology (2012-10-16)
S Belikov, C Öberg, T Jääskeläinen, V Rahkama, J J Palvimo, Ö Wrange
ABSTRACT
Prostate cancer growth depends on androgens. Synthetic antiandrogens are used in the cancer treatment. However, antiandrogens, such as bicalutamide (BIC), have a mixed agonist/antagonist activity. Here we compare the antiandrogenic capacity of BIC to a new antiandrogen, MDV3100 (MDV) or Enzalutamide™. By reconstitution of a hormone-regulated enhancer in Xenopus oocytes we show that both antagonists trigger the androgen receptor (AR) translocation to the nucleus, albeit with a reduced efficiency for MDV. Once in the nucleus, both AR-antagonist complexes can bind sequence specifically to DNA in vivo. The forkhead box transcription factor A (FoxA1) is a negative prognostic indicator for prostate cancer disease. FoxA1 expression presets the enhancer chromatin and makes the DNA more accessible for AR binding. In this context the BIC-AR antiandrogenic effect is seriously compromised as demonstrated by a significant chromatin remodeling and induction of a robust MMTV transcription whereas the MDV-AR complex displays a more persistent antagonistic character.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Bicalutamide (CDX), ≥98% (HPLC), powder