Merck
  • Small-molecule APOBEC3G DNA cytosine deaminase inhibitors based on a 4-amino-1,2,4-triazole-3-thiol scaffold.
Small-molecule APOBEC3G DNA cytosine deaminase inhibitors based on a 4-amino-1,2,4-triazole-3-thiol scaffold.
ChemMedChem (2012-11-28)
Margaret E Olson, Ming Li, Reuben S Harris, Daniel A Harki
ABSTRACT
APOBEC3G (A3G) is a single-stranded DNA cytosine deaminase that functions in innate immunity against retroviruses and retrotransposons. Although A3G can potently restrict Vif-deficient HIV-1 replication by catalyzing excessive levels of G→A hypermutation, sublethal levels of A3G-catalyzed mutation may contribute to the high level of HIV-1 fitness and its incurable prognosis. To chemically modulate A3G catalytic activity with the goal of decreasing the HIV-1 genomic mutation rate, we synthesized and biochemically evaluated a class of 4-amino-1,2,4-triazole-3-thiol small-molecule inhibitors identified by high-throughput screening. This class of compounds exhibits low-micromolar (3.9-8.2 μM) inhibitory potency and remarkable specificity for A3G versus the related cytosine deaminase, APOBEC3A. Chemical modification of inhibitors, A3G mutational screening, and thiol reactivity studies implicate C321, a residue proximal to the active site, as the critical A3G target for this class of molecules.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,2,4-Triazole, 98%