Merck
Types of polydrug use among Spanish adolescents.
Addictive behaviors (2012-12-21)
Sílvia Font-Mayolas, Maria Eugènia Gras, Natàlia Cebrián, Anna Salamó, Montserrat Planes, Mark J M Sullman
ABSTRACT
This study examined the types of polydrug use among Spanish adolescents. 1501 high school students (50.6% female) aged 12 to 17years old (mean age=14.03, SD=1.28), from public schools in the city of Girona (Catalonia, Spain), completed the survey. In the previous six months 20.9% of the Spanish adolescents used alcohol, 18.8% tobacco, 10.5% cannabis and 0.7% cocaine. Specifically, 28.6% of the sample (n=429; 29% males and 28.2% females) used at least one drug and 13.9% reported polydrug use (n=208; 12.6% males and 15.1% females). The present research also found that the most critical ages for starting polydrug use were 14 and 15years old. More than one quarter (27.9%) of the adolescent polydrug users were type A (tobacco and alcohol), 67.8% were type B (cannabis together with tobacco and/or alcohol) and 3.4% were type C (cannabis together with tobacco and/or alcohol, and cocaine). These results suggest that Spanish adolescents, particularly males, commence polydrug use at an earlier age than other European adolescents. Early preventative strategies and a multisubstance perspective are greatly needed in Spain to avoid the initiation of polydrug use or to prevent progress onto heavier drugs.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
(±)-MDMA solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-3,4-Methylenedioxymethamphetamine hydrochloride