Merck
Si-H activation in an iridium nitrido complex--a mechanistic and theoretical study.
Journal of the American Chemical Society (2013-02-26)
Daniel Sieh, Peter Burger
ABSTRACT
Si-H activation in triethyl- and triarylsilanes by a square-planar pyridine-diimine iridium complex with a terminal nitrido unit leads to the corresponding silyl amido complexes, which were unambiguously characterized by X-ray crystallography. Based on detailed combined kinetic and theoretical studies (DFT), direct addition of the Si-H bond to the iridium nitrido unit is proposed. The electronic propensities of the transition states for the Si-H activation were probed with a Hammett series of para-substituted triarylsilanes HSi(C6H5)2(4-C6H4-X). Based on the combination of experimental and theoretical studies, two independent pathways for this process are proposed, which point toward an ambiphilic propensity of the nitrido unit. Alternative pathways and the charge transfer in the transition states were also investigated. Furthermore, the barriers for the related H-H and C-H activation processes in dihydrogen and methane were analyzed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Piridina, ACS reagent, ≥99.0%
Sigma-Aldrich
Piridina, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Piridina, anhydrous, 99.8%
Sigma-Aldrich
Piridina, ReagentPlus®, ≥99%
Sigma-Aldrich
Piridina, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Piridina, ≥99%
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Piridina, biotech. grade, ≥99.9%
Supelco
Piridina, analytical standard
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Piridina, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol