Merck
  • Concentration-dependent inhibitory effects of baicalin on the metabolism of dextromethorphan, a dual probe of CYP2D and CYP3A, in rats.
Concentration-dependent inhibitory effects of baicalin on the metabolism of dextromethorphan, a dual probe of CYP2D and CYP3A, in rats.
Chemico-biological interactions (2013-03-06)
Xin Tian, Zhen-Yu Cheng, Jing He, Lin-Jing Jia, Hai-Ling Qiao
ABSTRACT
Baicalin has been shown to possess many pharmacological effects, including antiviral, antioxidant, anti-cancer and anti-inflammatory properties. In the current study, we reveal the inhibitory effects of baicalin on the metabolism of dextromethorphan (DXM), a dual probe substrate of CYP2D and CYP3A, in rats. Lineweaver-Burk plots demonstrated that baicalin inhibited the activities of CYP2D and CYP3A in a non-competitive manner in rat liver microsomes (RLMs). Concomitant administration of baicalin (0.90 g/kg, i.v.) and DXM (10 mg/kg, i.v.) increased the maximum drug concentration (C(max)) (37%) and the area under concentration-time curve (AUC) (42%) and decreased the clearance (CL) (27%) of DXM in a randomised, crossover study in rats (P < 0.01). The change in the AUC of DXM was significantly correlated with the C(max) and AUC of baicalin (P < 0.05). The inhibitory effects of multiple doses of baicalin (0.90 g/kg, i.v., 12 days) on the metabolism of DXM were similar to those observed following a single dose in rats. The activity of CYP3A in excised liver samples from rats following multiple baicalin treatment was significantly decreased compared to that of the control group (P < 0.05), whereas multiple doses of baicalin had no obvious effect on the activity of CYP2D. Taken together, these data demonstrate that baicalin inhibits the metabolism of DXM in a concentration-dependent manner in rats, possibly through inhibiting hepatic CYP2D and CYP3A activities.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Dextromethorphan HBr, certified reference material
Sigma-Aldrich
Baicalin, 95%
Sigma-Aldrich
Dextromethorphan hydrobromide monohydrate, ≥99% (TLC)
Supelco
Dextromethorphan solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dextromethorphan hydrobromide, meets USP testing specifications
Dextromethorphan hydrobromide, European Pharmacopoeia (EP) Reference Standard