Merck
Argan oil and postmenopausal Moroccan women: impact on the vitamin E profile.
Natural product communications (2013-03-12)
Hanae El Monfalouti, Zoubida Charrouf, Asma El Hamdouchi, Hanane Labraimi, Imane Chafchaouni-Moussaoui, Badreddine Kartah, Khalid El Kari, Yahya Bensouda, Abdelfettah Derouich, Sylvie Dodin, Clément Denhez, Dom Guillaume, Hassan Agnaou
ABSTRACT
Vitamin E supplements could be beneficial for postmenopausal women. To evaluate the effect of edible argan oil consumption on the antioxidant status of postmenopausal women, the vitamin E serum level of 151 menopausal women consuming either olive or argan oil was determined. Serum level of vitamin E was increased in the argan oil consumer group. Therefore, an argan oil-enriched diet can be recommended to help prevent some postmenopausal disorders.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Olive oil, highly refined, low acidity
Supelco
Olive oil, analytical standard
Sigma-Aldrich
Olive oil, tested according to Ph. Eur.