Merck
Diffusive hydrogen inter-cage migration in hydrogen and hydrogen-tetrahydrofuran clathrate hydrates.
The Journal of chemical physics (2013-03-15)
Huayu Cao, Niall J English, J M D MacElroy
ABSTRACT
Classical equilibrium molecular dynamics simulations have been performed to investigate the diffusive properties of inter-cage hydrogen migration in both pure hydrogen and mixed hydrogen-tetrahydrofuran sII hydrates at 0.05 kbar from 200 K and up to 250-260 K. For mixed H2-THF systems in which there is single H2 occupation of the small cage (labelled "1S1L"), we found that no H2 migration occurs. However, for more densely filled H2-THF and pure-H2 systems, in which there is more than single H2 occupation in the small cage, there is an onset of inter-cage H2 migration events from the small cages to neighbouring cavities at around 200 K. The mean square displacements of the hydrogen molecules were fitted to a mathematical model consisting of an anomalous term and a Fickian component, and nonlinear regression fitting was conducted to estimate long-time (inter-cage) diffusivities. An approximate Arrhenius temperature relationship for the diffusion coefficient was examined and an estimation of the hydrogen hopping energy barrier was calculated for each system.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tetraidrofurano, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetraidrofurano, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetraidrofurano, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetraidrofurano, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetraidrofurano, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Tetraidrofurano, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Supelco
Tetraidrofurano, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetraidrofurano, analytical standard
Supelco
Tetraidrofurano, Selectophore, ≥99.5%