Merck
Effect of temperature on toxicity of three pyrethroids to horn flies.
Medical and veterinary entomology (1989-10-01)
N C Hinkle, D C Sheppard, K Bondari, J F Butler
ABSTRACT
Predictive models describing best-fit regression equations for per cent mortality of horn flies as a function of temperature were determined for each of three pyrethroid insecticides (fenvalerate, flucythrinate and permethrin) over the temperature range 20-35 degrees C. Susceptible horn flies, Haematobia irritans (L.), were exposed to c. an LC70 dose of each pyrethroid using a residue-on-glass method. This technique used confined exposure in chambers with temperatures of 20, 25, 30 and 35 degrees C. Within this range, mortality was greatest at 25 degrees C with all three insecticides. Estimated temperature-mortality equations for each pyrethroid revealed different responses of horn flies to each of these insecticides. Horn flies exposed to flucythrinate demonstrated a linear mortality response that varied inversely with temperature. The response to permethrin was described by a quadratic equation, while the response to fenvalerate was best fitted by a cubic equation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Flucythrinate, PESTANAL®, analytical standard, mixture of stereo isomers