Merck
Headspace gas chromatography of stimulants in urine by in-column trifluoroacetyl derivatization method.
Journal of chromatography (1989-04-21)
H Tsuchihashi, K Nakajima, M Nishikawa, K Shiomi, S Takahashi
ABSTRACT
The analysis of stimulants in urine using a headspace gas chromatography system equipped with an in-column sample trifluoroacetylation unit was investigated. A 5-ml aliquot of urine containing stimulants was pipetted into a 20-ml autosampler vial together with 3.5 g of potassium carbonate. The vial was sealed and heated for 20 min at 80 degrees C, then 0.8 ml of the headspace gas and N-methylbis(trifluoroacetamide) gas were injected simultaneously into the gas chromatograph equipped with a flame ionization detector and a fused-silica capillary column (DB-1, 30 m x 0.32 mm I.D., film thickness 0.25 micron), with a gas-tight syringe. Calibration graphs prepared by the absolute calibration curve method showed good linearity over the concentration range of 0.04 to 50 micrograms/ml for methamphetamine hydrochloride and amphetamine sulphate. The detection limits were 0.03 micrograms/ml for both of these compounds.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥99.0% (GC)