- Sublethal effects of tetrachloro-1,2-benzoquinone--a component in bleachery effluents from pulp mills--on vertebral quality and physiological parameters in fourhorn sculpin.
Sublethal effects of tetrachloro-1,2-benzoquinone--a component in bleachery effluents from pulp mills--on vertebral quality and physiological parameters in fourhorn sculpin.
Ecotoxicology and environmental safety (1988-02-01)
B E Bengtsson, A Larsson, A Bengtsson, L Renberg
PMID3359957
ABSTRACT
The effects of tetrachloro-1,2-benzoquinone (TCQ), a component in bleached kraft mill effluents (BKME), on vertebral and physiological parameters were investigated in juvenile fourhorn sculpin, Myoxocephalus quadricornis L. After about 4.5 months of exposure to 0.1 and 0.5 mg TCQ/liter in 7% salinity brackish water, the fish demonstrated vertebral deformities, aberrant mechanical properties of the vertebrae, effects on white and red blood cell counts, enhanced activity of delta-aminolevulinic acid dehydratase in erythrocytes, and increased levels of ascorbic acid in the liver. The effects are discussed in relation to those previously observed in fish exposed to complex BKME in the field and the laboratory.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto