- Allergic contact dermatitis from 2-n-octyl-4-isothiazolin-3-one, a paint mildewcide.
Allergic contact dermatitis from 2-n-octyl-4-isothiazolin-3-one, a paint mildewcide.
Contact dermatitis (1983-11-01)
C G Mathias, K E Andersen, K Hamann
PMID6228380
ABSTRACT
Severe allergic contact dermatitis from a paint mildewcide, 2-n-octyl-4-isothiazolin-3-one, developed in a worker formulating latex paints within a paint manufacturing company, Guinea pig maximization testing demonstrated this to be a moderate sensitizer. Further cases of allergic contact dermatitis may be encountered as the use of this biocide increases in the market place.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto