Merck
  • Theoretical approach for radiosensitizers. Correlation between calculated electroaffinity and sensitization factors of nitro-compounds.
Theoretical approach for radiosensitizers. Correlation between calculated electroaffinity and sensitization factors of nitro-compounds.
International journal of radiation biology and related studies in physics, chemistry, and medicine (1981-06-01)
Y G Smeyers, A De Bueren, R Alcala, M V Alvarez
ABSTRACT
The electron affinity of eight well-known nitroaromatic and nitroheterocyclic radiosensitizers is computed resorting to a quantum mechanical approach. Four of the compounds studied were nitroimidazoles; but p-nitroacetophenone and some 5-nitrofuran derivatives were also investigated for comparison. A good correlation between the theoretical electroaffinity values and the radiosensitization efficiencies is found in the case of the nitroimidazoles, in which the pi-electron system appears to be limited to the aromatic ring, except for small side groups. In the case of molecules where the pi-electron system spreads out on the side-chain the theoretical procedure seems to fail.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
4′-Nitroacetophenone, 98%