Merck
  • Enhanced biodegradation and emulsification of crude oil and hyperproduction of biosurfactants by a gamma ray-induced mutant of Pseudomonas aeruginosa.
Enhanced biodegradation and emulsification of crude oil and hyperproduction of biosurfactants by a gamma ray-induced mutant of Pseudomonas aeruginosa.
Letters in applied microbiology (1995-09-01)
S Iqbal, Z M Khalid, K A Malik
ABSTRACT
A gamma ray-induced mutant of Pseudomonas aeruginosa strain S8, capable of hyperproduction of biosurfactant from hydrocarbons, was isolated and named as EBN-8. The mutant showed 3-4 times more hydrocarbon emulsification/conversion as compared to the parent when grown on Khaskheli crude oil in minimal medium. Enhanced biosurfactant production and hydrocarbon utilization by the mutant was also observed during growth on heptadecane in minimal medium as indicated by emulsion index and surface tension of cell-free culture broth. Using heptadecane as carbon and energy source, time course for the growth (cfu ml-1) and biosurfactant production were compared for both parent and mutant. These studies were carried out for 24 d at 30 +/- 2 degrees C and for 20 d at 37 degrees C. Growth of EBN-8 was much faster compared to the parent as well as being 2-3 times more hyperproductive.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Heptadecane, 99%
Supelco
Heptadecane, analytical standard