Merck
Cadmium ions inhibit procollagen C-proteinase and cupric ions inhibit procollagen N-proteinase.
Matrix biology : journal of the International Society for Matrix Biology (1994-03-01)
Y Hojima, B Behta, A M Romanic, D J Prockop
ABSTRACT
Procollagen C- and N-proteinases specifically cleave the C- and N-terminal extension propeptides of type I, II and III procollagen molecules. The collagen molecules generated by the enzymes self-assemble into collagen fibrils. We previously observed the inhibition of these enzymes purified from chick tendons by several divalent metals. Here the inhibitory effects of CdCl2, CuCl2, ZnCl2, NiCl2, CoCl2 and Hg(C2H3O2)2 have been studied in detail using crude or purified C- and N-proteinases from chick tendons and sterna. CdCl2 was a strong inhibitor of C-proteinases from both sources, and the inhibition was independent of enzyme purity (I50 = 10-16 microM). In contrast, CuCl2 and ZnCl2 were inhibitory only of purified C-proteinase. With the N-proteinase, CuCl2 was a strong inhibitor, and the inhibition was independent of the purity of the enzyme preparation used (I50 = 14-40 microM). On the other hand, CdCl2 was a moderate inhibitor, and ZnCl2 was a strong inhibitor only of the purified N-proteinase (I50 = 8-17 microM). NiCl2 inhibited crude and purified N-proteinase from sternum (I50 = 23-29 microM) but not from tendon. These results suggest, therefore, that the accumulation of some of these metals in the body may cause suppression of collagen fibril formation in tissues.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Mercury(II) acetate, ACS reagent, ≥98.0%
Sigma-Aldrich
Mercury(II) acetate, puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)