- Peripheral esterases in the rat: effects of classical inducers.
Peripheral esterases in the rat: effects of classical inducers.
Chemico-biological interactions (1993-06-01)
N W McCracken, P G Blain, F M Williams
PMID8343974
ABSTRACT
Liver microsomal paraoxonase, aryl esterase and fluazifop butyl esterase (carboxylesterase) were induced by pretreatment of rat with phenobarbitone but not by beta-naphthoflavone or clofibric acid. In the extrahepatic tissues lung cytosolicfluazifop butyl and phenylacetate esterase were induced.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto