Merck
  • Worm kinetics and serum IgE in hooded lister rats infected with the acanthocephalan Moniliformis moniliformis and the nematode Nippostrongylus brasiliensis.
Worm kinetics and serum IgE in hooded lister rats infected with the acanthocephalan Moniliformis moniliformis and the nematode Nippostrongylus brasiliensis.
International journal for parasitology (1996-11-01)
O Hindsbo, P A Anderson, P Lind
ABSTRACT
After infection with the intestinal helminths Moniliformis moniliformis or Nippostrongylus brasiliensis, worm-specific IgE first appeared in the serum rats between days 10 and 24 p.i., varying with host age, worm species and worm dose used. The rate of increase in specific IgE was comparable regardless of the worm species, infection dose or host age and a peak response was observed about 1 month after the sera turned positive. In the M. moniliformis infections, these events took place long before the beginning of worm expulsion on day 63 in high-dose (50 worms) infections, and potentiation of heterologous IgE was not observed. In contrast, IgE stimulation by N. brasiliensis infections was detected as potentiation of anti-ovalbumin IgE, anti-M. moniliformis IgE and total IgE. Most of the total IgE in the serum of M. moniliformis-infected rats was likely to be the worm-specific IgE. Anthelminthic removal of M. moniliformis revealed that the presence of residual worms was necessary to maintain worm-specific IgE production.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Oxyclozanide, VETRANAL®, analytical standard