Merck
Participation of biological membranes in colloidal gold transformation by Micrococcus luteus cells.
Membrane & cell biology (1997-01-01)
L A Levchenko, A P Sadkov, S A Marakushev, N V Lariontseva
ABSTRACT
Methods of biochemical and physicochemical analysis were applied for studying the role of Micrococcus luteus cell structure in gold accumulation. It was shown that membrane proteins were the main factors in this process. Moreover, different quinones played the main role in the redox transformation of gold.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%