Merck
  • Physicochemical and immunological studies of the N-terminal domain of the Torpedo acetylcholine receptor alpha-subunit expressed in Escherichia coli.
Physicochemical and immunological studies of the N-terminal domain of the Torpedo acetylcholine receptor alpha-subunit expressed in Escherichia coli.
European journal of biochemistry (1999-01-23)
T Alexeev, A Krivoshein, A Shevalier, I Kudelina, O Telyakova, A Vincent, Y Utkin, F Hucho, V Tsetlin
ABSTRACT
The nicotinic acetylcholine receptor (AChR) from the electric organ of Torpedo species is an oligomeric protein composed of alpha2 beta gamma delta subunits. Although much is known about its tertiary and quaternary structure, the conformation of the large extracellular domains of each of the subunits has not been analysed in detail. In order to obtain information about the spatial structure of the extracellular domain, we have expressed the N-terminal fragment 1-209 of the Torpedo californica AChR alpha-subunit in Escherichia coli. Two vectors coding for a (His)6 tag, either preceding or following the 1-209 sequence, were used and the recombinant proteins obtained (designated alpha1-209pET and alpha1-209pQE, respectively) were purified by affinity chromatography on a Ni2+-agarose column. The chemical structure of both proteins was verified by Edman degradation and mass spectrometry. The proteins were soluble in aqueous buffers but to make possible a comparison with the whole AChR or its isolated subunits, the recombinant proteins were analyzed both in aqueous solution and with the addition of detergents. The two proteins bound [125I]alpha-bungarotoxin with equal potency (KD approximately 130 nm, Bmax approximately 10 nmol.mg-1). Both were shown to interact with several monoclonal antibodies raised against purified Torpedo AChR. The circular dichroism (CD) spectra of the two proteins in aqueous solution revealed predominantly beta-structure (50-56%), the fraction of alpha-helices amounting to 32-35%. Nonionic (beta-octylglucoside) and zwitterionic (CHAPS) detergents did not appreciably change the CD spectra, while the addition of SDS or trifluoroethanol decreased the percentage of beta-structure or increased the alpha-helicity, respectively. The predominance of beta-structure is in accord with recent data on the N-terminal domain of the mouse muscle AChR alpha-subunit expressed in the mammalian cells [West et al. (1997) J. Biol. Chem. 272, 25 468]. Thus, expression in E. coli provides milligram amounts of the protein that retains several structural characteristics of the N-terminal domain of the Torpedo AChR alpha-subunit and appears to share with the latter a similar secondary structure. The expression of recombinant polypeptides representing functional domains of the AChR provides an essential first step towards a more detailed structural analysis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Decamethonium bromide, crystalline
Sigma-Aldrich
Decamethonium bromide, Ion-Pair reagent, ≥99%