Merck
Phenylene ethynylene diazonium salts as potential self-assembling molecular devices.
Organic letters (2001-03-30)
D V Kosynkin, J M Tour
ABSTRACT
[reaction: see text]. Functionalized diazonium salts for molecular electronic devices are prepared by the reaction of the corresponding anilines with NOBF4 in sulfolane-acetonitrile solvent.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sulfolano, 99%
Supelco
Sulfolano, analytical standard