Merck
Optimized dyeing conditions of immunoprotein with reactive dye Procion Blue MX-7RX.
Analytical biochemistry (2007-01-02)
Shwu-Jinng Wang, Hsiu-Min Lin, Xin-Ren Wang, Kuang-Pin Hsiung, Yung-Chuan Liu
ABSTRACT
For investigating the feasibility of using reactive dyes as an immunoassay marker, a dichlorine triazine dye, Procion Blue MX-7RX, was employed to stain the antibody against human serum albumin (anti-HSA). With the color intensity revealed in the immunochromatographic test strip as the objective variables, the optimal dyeing conditions were found as follows: pH 11.4, temperature 35.7 degrees C, molar ratio 188 (mol dye/mol antibody), and reaction time 45.6 min. The dyed-anti-HSA revealed a maximal color intensity of 8738 without apparent loss of antigen binding affinity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Reactive Blue 4, Dye content 35 %