Merck
  • Validated HPTLC methods for the determination of salicin in Salix sp. and of harpagoside in Harpagophytum [corrected] procumbens.
Validated HPTLC methods for the determination of salicin in Salix sp. and of harpagoside in Harpagophytum [corrected] procumbens.
Journal of pharmaceutical and biomedical analysis (2008-07-08)
Steffen Wagner, Abraham Ureña, Eike Reich, Irmgard Merfort
ABSTRACT
Preparations of Harpagophytum procumbens and of Salix species are successfully used for the treatment of degenerative rheumatism and painful arthrosis. For the quality control of both drugs, rapid methods of quantification are desirable. Here we report the development of two HPTLC methods enabling the determination of harpagoside in Harpagophyti radix and of salicin in Salicis cortex. We focused on a standardized methodology and thorough validation including two laboratories. The methods allow the analyses of up to 16 samples in parallel demonstrating the proposed methods as very rapid and cost efficient.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Harpagoside, analytical standard
Harpagoside, primary reference standard