Merck
  • Transfer of oligosaccharide from oligosaccharide pyrophosphoryl dolichol to endogenous acceptor proteins in human breast malignant and normal tissues.
Transfer of oligosaccharide from oligosaccharide pyrophosphoryl dolichol to endogenous acceptor proteins in human breast malignant and normal tissues.
Experientia (1985-07-15)
A R Santa Cruz, A Baldi
ABSTRACT
We have prepared dolichylpyrophosphoryl-[14C]-oligosaccharide (Dol-PP-oligosaccharide) from calf thyroid. Microsomal fractions from human breast tissues catalyzed the transfer of labeled oligosaccharide to endogenous acceptor proteins. Malignant tumors showed higher activity of the oligosaccharide transferring enzyme than normal tissue. With kojibiose (Kj), an inhibitor of (Glc3)-glucosidase, an increase in the radioactivity associated with glycoprotein was observed.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Kojibiose, ≥98% (HPLC)