Merck
  • Gastric submucosal microdialysis: a method to study gastrin- and food-evoked mobilization of ECL-cell histamine in conscious rats.
Gastric submucosal microdialysis: a method to study gastrin- and food-evoked mobilization of ECL-cell histamine in conscious rats.
Regulatory peptides (2000-02-15)
M Kitano, P Norlén, R Håkanson
ABSTRACT
Rat stomach ECL cells are rich in histamine and chromogranin A-derived peptides, such as pancreastatin. Gastrin causes the parietal cells to secrete acid by flooding them with histamine from the ECL cells. In the past, gastric histamine release has been studied using anaesthetized, surgically manipulated animals or isolated gastric mucosa, glands or ECL cells. We monitored gastric histamine mobilization in intact conscious rats by subjecting them to gastric submucosal microdialysis. A microdialysis probe was implanted into the submucosa of the acid-producing part of the stomach (day 1). The rats had access to food and water or were deprived of food (48 h), starting on day 2 after implantation of the probe. On day 4, the rats received food or gastrin (intravenous infusion), and sampling of microdialysate commenced. Samples (flow rate 1.2 microl min(-1)) were collected every 20 or 60 min, and the histamine and pancreastatin concentrations were determined. The serum gastrin concentration was determined in tail vein blood. Exogenous gastrin (4-h infusion) raised microdialysate histamine and pancreastatin dose-dependently. This effect was prevented by gastrin receptor blockade (YM022). Depletion of ECL-cell histamine by alpha-fluoromethylhistidine, an irreversible inhibitor of the histamine-forming enzyme, suppressed the gastrin-evoked release of histamine but not that of pancreastatin. Fasting lowered serum gastrin and microdialysate histamine by 50%, while refeeding raised serum gastrin and microdialysate histamine and pancreastatin 3-fold. We conclude that histamine mobilized by gastrin and food intake derives from ECL cells because: 1) Histamine and pancreastatin were released concomitantly, 2) histamine mobilization following gastrin or food intake was prevented by gastrin receptor blockade, and 3) mobilization of histamine (but not pancreastatin) was abolished by alpha-fluoromethylhistidine. Hence, gastric submucosal microdialysis allows us to monitor the mobilization of ECL-cell histamine in intact conscious rats under various experimental conditions not previously accessible to study. While gastrin receptor blockade lowered post-prandial release of ECL-cell histamine by about 80%, unilateral vagotomy reduced post-prandial mobilization of ECL-cell histamine by about 50%. Hence, both gastrin and vagal excitation contribute to the post-prandial release of ECL-cell histamine.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
YM 022, ≥98% (HPLC)