Merck
Effects of subinhibitory concentrations of nitroxoline on the surface properties of Escherichia coli.
Folia microbiologica (2001-08-15)
H Latrache, P Bourlioux, M Karroua, H Zahir, A Hakkou
ABSTRACT
Nitroxoline (5-nitro-8-quinolinol; NIQ) at subinhibitory concentrations (sub-MIC) decreased the adherence of uropathogenic Escherichia coli to catheter surface and significantly enhanced cell surface hydrophobicity. The surface hydrophobicity increased in the presence of sub-MIC of NIQ and also in an excess of Mg2+. The effect of NIQ on the cell surface was not related to the bacteriostatic effect of this agent. The increase in nitrogen and decrease in phosphate content in the cell surface was found in the presence of NIQ. NIQ did not inhibit the expression of fimbriae.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
8-Hydroxy-5-nitroquinoline, 96%
Prezzi e disponibilità al momento non sono disponibili