Merck
  • The actions of a D-1 agonist in MPTP treated primates show dependence on both D-1 and D-2 receptor function and tolerance on repeated administration.
The actions of a D-1 agonist in MPTP treated primates show dependence on both D-1 and D-2 receptor function and tolerance on repeated administration.
Journal of neural transmission (Vienna, Austria : 1996) (2002-06-22)
L A Smith, M J Jackson, G Al-Barghouthy, P Jenner
ABSTRACT
The potent and long acting D-1 receptor agonist, A-77636 reverses motor deficits in MPTP treated common marmosets following subcutaneous or oral administration. We now explore the effects of acute versus repeated administration of A-77636 and the relative roles of D-1 and D-2 receptor involvement in its antiparkinsonian actions. Acute oral administration (0.18-9.0 mg/kg) or subcutaneous administration (0.036-1.08 mg/kg) of A-77636 produced well co-ordinated bouts of discontinuous locomotor activity. There was no evidence of repetitive or stereotyped movements. However, oral administration of A-77636 (1.44 mg/kg) on alternate days for 10 days produced tolerance to the increase in locomotor activity and improvement of disability. Pretreatment with the D-1 antagonist SCH 23390 (0.625, 2.5 or 5.0 mg/kg, intraperitoneally), produced a dose dependent reduction in locomotor activity and antagonised the reduction in disability scores following administration of A-77636 (0.36 mg/kg, subcutaneously). The inhibitory effects of SCH 23390 lasted for some 2-5 hours after which the activity of A-77636 was gradually restored. Unexpectedly, pretreatment with the D-2 antagonist raclopride (1.25, 5.0 or 20.0 mg/kg, intraperitoneally), dramatically diminished the antiparkinsonian effects of A-77636 (0.36 mg/kg, subcutaneously) in a dose dependent manner. The dependence of the antiparkinsonian activity of A-77636 on intact D-2 receptor function, suggests a need for endogenous D-2 receptor tone to express D-1 mediated locomotor activity.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
A-77636 hydrochloride hydrate, ≥98% (HPLC), solid