Merck
  • Impairment of carbamazepine-10, 11-epoxide elimination by valnoctamide, a valpromide isomer, in healthy subjects.
Impairment of carbamazepine-10, 11-epoxide elimination by valnoctamide, a valpromide isomer, in healthy subjects.
British journal of clinical pharmacology (1992-07-01)
F Pisani, A Fazio, C Artesi, G Oteri, E Spina, T Tomson, E Perucca
ABSTRACT
The effect of the valpromide isomer valnoctamide (VCD, 200 mg three times daily for 8 days), an over-the-counter tranquillizer, on the elimination kinetics of a single oral dose of carbamazepine-10, 11-epoxide (CBZ-E, 100 mg) was investigated in healthy subjects. During VCD treatment, the half-life of CBZ-E was prolonged significantly compared with control (19.7 +/- 6.7 h vs 6.9 +/- 2.0 h, means +/- s.d., P less than 0.01), and its oral clearance decreased four-fold (from 109.6 +/- 30.7 to 28.8 +/- 11.1 ml h-1 kg-1, P less than 0.01). These findings indicate that VCM, like valpromide, strongly inhibits epoxide hydrolase in vivo.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Valnoctamide, ≥98% (NMR)