Merck
  • o-Nitrotoluene-induced large intestinal tumors in B6C3F1 mice model human colon cancer in their molecular pathogenesis.
o-Nitrotoluene-induced large intestinal tumors in B6C3F1 mice model human colon cancer in their molecular pathogenesis.
Carcinogenesis (2003-12-23)
R C Sills, H L Hong, G Flake, C Moomaw, N Clayton, G A Boorman, J Dunnick, T R Devereux
ABSTRACT
In the previous 500 2-year chemical bioassays within the National Toxicology Program, large intestinal tumors (cecal carcinomas) related to chemical exposure have not been observed in B6C3F1 mice. The recently completed o-nitrotoluene study provided the first cecal tumor response and an opportunity to evaluate the morphology and molecular profile of oncogenes and tumor suppressor genes that are relevant to humans. Morphologically, the carcinomas were gland-forming tumors lined by tall columnar epithelial cells that were positive for cytokeratin 20 and negative for cytokeratin 7. Using immunohistochemistry beta-catenin (encoded by Catnb) protein accumulation was detected in 80% (8/10) of the cecal carcinomas, while increased cyclin D1 and p53 protein expression was detected in 73% (8/11), respectively. There was no difference in adenomatous polyposis protein expression between normal colon and cecal carcinomas. All tumors examined exhibited mutations in exon 2 (corresponds to exon 3 in humans) in the Catnb gene. Mutations in p53 were identified in nine of 11 carcinomas, and all were in exon 7. Analysis of the K-ras gene revealed mutations in 82% (9/11) of carcinomas; all had specific G --> T transversions (Gly --> Val) at codons 10 or 12. The alterations in cancer genes and proteins found in the mouse large intestinal tumors included mutations that activate signal transduction pathways (K-ras and Catnb) and changes that disrupt the cell-cycle and bypass G(1) arrest (p53, cyclin D1). These alterations, which are hallmarks of human colon cancer, probably contributed to the pathogenesis of the large intestinal carcinomas in mice following o-nitrotoluene exposure.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Nitrotoluene, ≥99%