Merck
[Chemical components of volatiles form withered black poplar leaves with different physiological age].
Ying yong sheng tai xue bao = The journal of applied ecology (2006-01-21)
Xianru Guo, Guohui Yuan, Jinwei Jiang, Meihao Luo, Jisheng Ma
ABSTRACT
By the methods of steam distillation and GC-MS, this paper analyzed the chemical components of the volatiles from withered black poplar leaves. The main components of the volatiles from young leaves were(Z)-3-hexenol (44.81% ),4-methyl-1-pentanol (21.85%) and 2-hydroxy-benzaldehyde (15.19%), those from matured leaves were(Z)-3-hexenol (28.71%) and 2-hydroxy-benzaldehyde (10.35%), while in the volatiles from senescent leaves, the main components were 2-hydroxy-benzaldehyde (28.81%) and benzyl alcohol (15.06%). The (Z)-3-hexenol content in the volatiles was evidently decreased with increasing leaf age, while the species and contents of aromatic compounds were in adverse.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
α-Isobutylphenethyl alcohol, 98%