Merck
  • The ACAT inhibitor avasimibe increases the fractional clearance rate of postprandial triglyceride-rich lipoproteins in miniature pigs.
The ACAT inhibitor avasimibe increases the fractional clearance rate of postprandial triglyceride-rich lipoproteins in miniature pigs.
Biochimica et biophysica acta (2006-01-24)
John R Burnett, Dawn E Telford, P Hugh R Barrett, Murray W Huff
ABSTRACT
Previously, we have shown, in vivo, that the acyl coenzyme A: cholesterol acyltransferase (ACAT) inhibitor avasimibe decreases hepatic apolipoprotein (apo) B secretion into plasma. To test the hypothesis that avasimibe modulates postprandial triglyceride-rich lipoprotein (TRL) metabolism in vivo, an oral fat load (2 g fat/kg) containing retinol was given to 9 control miniature pigs and to 9 animals after 28 days treatment with avasimibe (10 mg/kg/day, n=5; 25 mg/kg/day, n=4). The kinetic parameters for plasma retinyl palmitate (RP) metabolism were determined by multi-compartmental modeling using SAAM II. Avasimibe decreased the 2-h TRL (d<1.006 g/mL; S(f)>20) triglyceride concentrations by 34%. The TRL triglyceride 0-12 h area under the curve (AUC) was decreased by 21%. In contrast, avasimibe had no effect on peak TRL RP concentrations, time to peak, or its rate of appearance into plasma, however, the TRL RP 0-12 h AUC was decreased by 17%. Analysis of the RP kinetic parameters revealed that the TRL fractional clearance rate (FCR) was increased 1.4-fold with avasimibe. The TRL RP FCR was negatively correlated with very low density lipoprotein (VLDL) apoB production rate measured in the fasting state (r=-0.504). No significant changes in total intestinal lipid concentrations were observed. Thus, although avasimibe had no effect on intestinal TRL secretion, plasma TRL clearance was significantly increased; an effect that may relate to a decreased competition with hepatic VLDL for removal processes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Avasimibe, ≥98% (HPLC)