Merck
Arachidonyl trifluoromethyl ketone is neuroprotective after spinal cord injury.
Journal of neurotrauma (2009-04-18)
Wenlong Huang, Amar Bhavsar, Rachael E Ward, Jodie C E Hall, John V Priestley, Adina T Michael-Titus
ABSTRACT
In spinal cord injury (SCI), neuronal and oligodendroglial loss occurs as a result of the initial trauma and the secondary damage that is triggered by excitotoxicity, free radicals, and inflammation. There is evidence that SCI ellicits increased cytosolic phospholipase A(2) (cPLA(2)) activity. The cleavage of phospholipids by cPLA(2) leads to release of fatty acids, and in particular arachidonic acid (AA), the metabolites of which have been associated with increased inflammation and oxidative stress. The aim of our study was to investigate whether the inhibition of cPLA(2) following SCI leads to tissue protection and an improved functional outcome. Adult rats received compression SCI and 30 min after injury they were treated intravenously with either saline or the cPLA(2) inhibitor arachidonyl trifluoromethyl ketone (AACOCF3) (7.13 mg/kg). The animals were sacrificed at 7 days post-injury and the lesioned tissue was labeled using markers for neurons, oligodendrocytes, and macrophages/activated microglia. We also assessed locomotor recovery using the Basso-Beattie-Bresnahan (BBB) score. The number of surviving neurons and oligodendrocytes was significantly increased in animals treated with the cPLA(2) inhibitor compared to saline controls. The behavioral analysis mirrored the neuroprotective effects and showed that the inhibitor-treated group had better locomotor recovery compared to saline controls. Our results show that AACOCF3 has neuroprotective potential, and support the idea that cPLA(2) is critically involved in acute spinal injury.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Arachidonyl trifluoromethyl ketone, oil, ≥97% (NMR)