Merck
  • Investigation into UDP-glucuronosyltransferase (UGT) enzyme kinetics of imidazole- and triazole-containing antifungal drugs in human liver microsomes and recombinant UGT enzymes.
Investigation into UDP-glucuronosyltransferase (UGT) enzyme kinetics of imidazole- and triazole-containing antifungal drugs in human liver microsomes and recombinant UGT enzymes.
Drug metabolism and disposition: the biological fate of chemicals (2010-03-23)
Karine Bourcier, Ruth Hyland, Sarah Kempshall, Russell Jones, Jacqueline Maximilien, Nicola Irvine, Barry Jones
ABSTRACT
Imidazoles and triazoles represent major classes of antifungal azole derivatives. With respect to UDP-glucuronosyltransferase (UGT) enzymes, the drug metabolism focus has mainly concentrated on their inhibitory effects with little known about azoles as substrates for UGTs. N-Glucuronide metabolites of the imidazole antifungals, tioconazole and croconazole, have been reported, but there are currently no reports of N-glucuronidation of triazole antifungal agents. In this study, evidence for glucuronidation of azole-containing compounds was studied in human liver microsomes (HLM). When a glucuronide metabolite was identified, azoles were incubated in 12 recombinant UGT (rUGT) enzymes, and enzyme kinetics were determined for the UGT with the most intense glucuronide peak. Six imidazole antifungals, three triazoles, and the benzodiazepine alprazolam (triazole) were evaluated in this study. All compounds investigated were identified as substrates of UGT. UGT1A4 was the main enzyme involved in the metabolism of all compounds except for fluconazole, which was mainly metabolized by UGT2B7, probably mediating its O-glucuronide metabolism. UGT1A3 was also found to be involved in the metabolism of all imidazoles but not triazoles. In both HLM and rUGT K(m) values were lower for imidazoles (14.8-144 microM) than for triazoles (158-3037 microM), with the exception of itraconazole (8.4 microM). All of the imidazoles studied inhibited their own metabolism at high substrate concentrations. In terms of UGT1A4 metabolism, itraconazole showed kinetic features characteristic of imidazole rather than triazole antifungals. This behavior is attributed to the physicochemical properties of itraconazole that are similar to those of imidazoles in terms of clogP.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Tioconazole, 97%